Learn More
ZNF287 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 22162-1-AP-150UL
 View more versions of this product
Product Code. 16881695
  • 4652.90 SEK / 150µL
This item is not returnable. View return policy
Description
Description
This gene encodes a member of the krueppel family of zinc finger proteins, suggesting a role as a transcription factor. Its specific function has not been determined. This gene is located near the Smith-Magenis syndrome region on chromosome 17.
Specifications
Specifications
ZNF287
Polyclonal
Unconjugated
ZNF287
zinc finger protein 287, ZKSCAN13, ZNF287
Rabbit
Antigen Affinity Chromatography
RUO
57336
-20°C
Liquid
Western Blot
0.24 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9HBT7
ZNF287
ZNF287 Fusion Protein Ag17814
150 μL
Primary
Human
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
ZNF287 Rabbit anti-Human, Polyclonal, Proteintech > 150 μL; Unconjugated