missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ ZNF313 Recombinant Protein
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
Description
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Specifications
Specifications
Accession Number | AAH13695 |
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer |
Gene ID (Entrez) | 55905 |
Molecular Weight (g/mol) | 50.82 |
Name | ZNF313 (Human) Recombinant Protein (P01) |
pH Range | 8 |
Preparation Method | In vitro wheat germ expression system |
Purification Method | Glutathione Sepharose 4 Fast Flow |
Quality Control Testing | 12.5% SDS-PAGE Stained with Coomassie Blue. |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Abnova™ ZNF313 Recombinant Protein >
Spot an opportunity for improvement?Share a Content Correction