Learn More
ZNF346 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 20794-1-AP-20UL
 View more versions of this product
Product Code. 16890365
  • 1645.54 SEK / 20µL
Description
Probable transcriptional activator which may play a role in neuronal differentiation. Able to bind DNA and activate expression in vitro.
Specifications
Specifications
ZNF346
Polyclonal
Unconjugated
ZNF346
JAZ, Zfp346, zinc finger protein 346, ZNF346
Rabbit
Antigen Affinity Chromatography
RUO
23567, 26919, 306765
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin)
0.39 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9R0B7, Q9UL40
Zfp346, Znf346
ZNF346 Fusion Protein Ag14585
20 μL
Primary
Human, Rat, Mouse
Antibody
IgG
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
ZNF346 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated