Learn More
ZNF346 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20794-1-AP-150UL
This item is not returnable.
View return policy
Description
Probable transcriptional activator which may play a role in neuronal differentiation. Able to bind DNA and activate expression in vitro.Specifications
ZNF346 | |
Polyclonal | |
Unconjugated | |
ZNF346 | |
JAZ, Zfp346, zinc finger protein 346, ZNF346 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23567, 26919, 306765 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.39 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9R0B7, Q9UL40 | |
Zfp346, Znf346 | |
ZNF346 Fusion Protein Ag14585 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Product Title
ZNF346 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction