Learn More
ZNF346 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 20794-1-AP-150UL
 View more versions of this product
Product Code. 16880365
  • 4652.90 SEK / 150µL
This item is not returnable. View return policy
Description
Description
Probable transcriptional activator which may play a role in neuronal differentiation. Able to bind DNA and activate expression in vitro.
Specifications
Specifications
ZNF346
Polyclonal
Unconjugated
ZNF346
JAZ, Zfp346, zinc finger protein 346, ZNF346
Rabbit
Antigen Affinity Chromatography
RUO
23567, 26919, 306765
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.39 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9R0B7, Q9UL40
Zfp346, Znf346
ZNF346 Fusion Protein Ag14585
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
ZNF346 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated