Learn More
ZNF385D Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 20806-1-AP-20UL
 View more versions of this product
Product Code. 16890385
  • 1645.54 SEK / 20µL
This item is not returnable. View return policy
Description
Description
ZNF385D gene ontology annotations related to this gene include nucleic acid binding; zinc ion binding.
Specifications
Specifications
ZNF385D
Polyclonal
Unconjugated
ZNF385D
zinc finger protein 385D, Zinc finger protein 659, ZNF385D, ZNF659
Rabbit
Antigen Affinity Chromatography
RUO
79750
-20°C
Liquid
Western Blot
0.34 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9H6B1
ZNF385D
ZNF385D Fusion Protein Ag14746
20 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
ZNF385D Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 20 μL; Unconjugated