missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF497 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25178-1-AP-20UL
This item is not returnable.
View return policy
Description
May be involved in transcriptional regulation.Specifications
ZNF497 | |
Polyclonal | |
Unconjugated | |
ZNF497 | |
zinc finger protein 497, ZNF497 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
162968 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.45 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q6ZNH5 | |
ZNF497 | |
ZNF497 Fusion Protein Ag18114 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
ZNF497 Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction