missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ ZNF514 (Human) Recombinant Protein

Human ZNF514 full-length ORF ( NP_116177.1, 1 a.a. - 400 a.a.) recombinant protein with GST-tag at N-terminal.

3860.70 SEK - 5854.80 SEK

Specifications

Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Gene ID (Entrez) 84874
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16146133
View Documents Promotion Details
Abnova™
H00084874-P01.10UG
10 μg
3860.70 SEK
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 07-03-2022
Please to purchase this item. Need a web account? Register with us today!
16156133
View Documents Promotion Details
Abnova™
H00084874-P01.25UG
25 μg
5854.80 SEK
25µg
View alternative products
Save Up to 
»
Estimated Shipment: 07-03-2022
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Sequence: MTFEDVAVEFSQWEWGQLNPAQKDLYREVMLENFRNLAILGLLVSKPYVICQLEEGGEPFMVEREISTGAHSDWKRRSKSKESMPSWGISKEELFQVVSVEKHIQDVLQFSKLKAACGCDGQLEMQQIKQERHLKQMSTIHKSATTLSRDYKWNGFGRSLGLRSVLVNQHSILMGEGSYKCDTEFRQTLGGNNSQRTHPEKKSCKCNECGKSFHFQSELRRHQRCHTGEKPYECSDCGRAFGHISSLIKHQRTHTGEKPYECSECGRAFSQSSSLVLHYRFHTGEKPYKCNECGRAFGHTSSLIKHQRTHTGEKPYECRECGRTFSQSSSLIVHYRFHTGEKPYKCNKCGRAFSQSSSLTQHYRFHTGEKPYKCNECGRAFAHTASLIKHQRSHAGKKTL
Specifications

Specifications

Store at -80°C. Aliquot to avoid repeated freezing and thawing.
84874
SDS
Product Certifications