Learn More
ZNF593 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 19426-1-AP-150UL
 View more versions of this product
Product Code. 16888984
  • 4652.90 SEK / 150µL
This item is not returnable. View return policy
Description
Description
Negatively modulates the DNA binding activity of Oct-2 and therefore its transcriptional regulatory activity. Could act either by binding to DNA octamer or by interacting with Oct-2. May also be a modulator of other octamer-binding proteins.
Specifications
Specifications
ZNF593
Polyclonal
Unconjugated
ZNF593
zinc finger protein 593, Zinc finger protein T86, ZNF593, ZT86
Rabbit
Antigen Affinity Chromatography
RUO
51042, 68040
-20°C
Liquid
Western Blot
0.33 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O00488, Q9DB42
ZNF593
ZNF593 Fusion Protein Ag13758
150 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
ZNF593 Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 150 μL; Unconjugated