missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF674 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24747-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
This gene encodes a zinc finger protein with an N-terminal Kruppel-associated box-containing (KRAB) domain and 11 Kruppel-type C2H2 zinc finger domains. Like other zinc finger proteins, this gene may function as a transcription factor. This gene resides on an area of chromosome X that has been implicated in nonsyndromic X-linked mental retardation. Alternative splicing results in multiple transcript variants encoding different isoforms.Specifications
ZNF674 | |
Polyclonal | |
Unconjugated | |
ZNF674 | |
MRX92, zinc finger family member 674, Zinc finger protein 674, ZNF673B, ZNF674 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
641339 | |
-20°C | |
Liquid |
Western Blot | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q2M3X9 | |
ZNF674 | |
ZNF674 Fusion Protein Ag21576 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |