missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF674 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | ZNF674 |
---|---|
Concentration | 0.3 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16883235
|
Proteintech
24747-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16873235
|
Proteintech
24747-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This gene encodes a zinc finger protein with an N-terminal Kruppel-associated box-containing (KRAB) domain and 11 Kruppel-type C2H2 zinc finger domains. Like other zinc finger proteins, this gene may function as a transcription factor. This gene resides on an area of chromosome X that has been implicated in nonsyndromic X-linked mental retardation. Alternative splicing results in multiple transcript variants encoding different isoforms.Specifications
ZNF674 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q2M3X9 | |
641339 | |
ZNF674 Fusion Protein Ag21576 | |
Primary | |
-20°C | |
ZNF674 |
0.3 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
MRX92, zinc finger family member 674, Zinc finger protein 674, ZNF673B, ZNF674 | |
ZNF674 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
ZNF674 Rabbit anti-Human, Polyclonal, Proteintech