missing translation for 'onlineSavingsMsg'
Learn More

ZNF747 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

1645.54 SEK - 4652.90 SEK

Specifications

Antigen ZNF747
Concentration 0.18 mg/mL
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16899364
View Documents
Proteintech
19901-1-AP-20UL
20 ÎĽL N/A
N/A
16889364
View Documents
Proteintech
19901-1-AP-150UL
150 ÎĽL N/A
N/A
Description

Description

ZNF747 gene ontology annotations related to this gene include regulation of transcription, DNA-templated.
Specifications

Specifications

ZNF747
Western Blot
Unconjugated
Rabbit
Human
Q9BV97
65988
ZNF747 Fusion Protein Ag13821
Primary
-20°C
ZNF747
0.18 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
zinc finger protein 747, ZNF747
ZNF747
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
ZNF747 Rabbit anti-Human, Polyclonal, Proteintech

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.