Learn More
ZnT3 Mouse anti-Human, Unconjugated, Clone: 1B9B1, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67262-1-IG-20UL
 View more versions of this product
Product Code. 16889785
  • 1645.54 SEK / 20µL
Description
The ZnT3 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ZnT3. This antibody recognizes human, pig, rat antigen. The ZnT3 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
ZnT3
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Rat, Pig
Western Blot, ELISA
1B9B1
Unconjugated
20 μL
7781
IgG2b
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
ZnT3 Mouse anti-Human, Unconjugated, Clone: 1B9B1, Proteintech > 20 μL