Learn More
ZnT3 Mouse anti-Human, Unconjugated, Clone: 1B9B1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67262-1-IG-150UL
Description
The ZnT3 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ZnT3. This antibody recognizes human, pig, rat antigen. The ZnT3 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
ZnT3 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human, Rat, Pig |
Western Blot, ELISA | |
1B9B1 | |
Unconjugated | |
150 μL | |
7781 | |
IgG2b |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.