Learn More
ZW10 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24561-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
This gene encodes a protein that is one of many involved in mechanisms to ensure proper chromosome segregation during cell division.Specifications
ZW10 | |
Polyclonal | |
Unconjugated | |
ZW10 | |
HZW10, KNTC1AP, ZW10 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
9183 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.43 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O43264 | |
ZW10 | |
ZW10 Fusion Protein Ag19138 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |