missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZW10 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | ZW10 |
---|---|
Concentration | 0.43 mg/mL |
Applications | Western Blot, Immunoprecipitation |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16823045
|
Proteintech
24561-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16813045
|
Proteintech
24561-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a protein that is one of many involved in mechanisms to ensure proper chromosome segregation during cell division.Specifications
ZW10 | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Human | |
O43264 | |
9183 | |
ZW10 Fusion Protein Ag19138 | |
Primary | |
-20°C | |
ZW10 |
0.43 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
HZW10, KNTC1AP, ZW10 | |
ZW10 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |