Learn More
ZWINT Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12282-2-AP-20UL
 View more versions of this product
Product Code. 16879903
  • 1645.54 SEK / 20µL
This item is not returnable. View return policy
Description
Description
This gene encodes a protein that is clearly involved in kinetochore function although an exact role is not known. It interacts with ZW10, another kinetochore protein, possibly regulating the association between ZW10 and kinetochores. The encoded protein localizes to prophase kinetochores before ZW10 does and it remains detectable on the kinetochore until late anaphase. It has a uniform distribution in the cytoplasm of interphase cells. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications
Specifications
ZWINT
Polyclonal
Unconjugated
ZWINT
HZwint 1, KNTC2AP, ZW10 interacting protein 1, ZW10 interactor, ZWINT, Zwint 1, ZWINT1
Rabbit
Antigen Affinity Chromatography
RUO
11130
-20°C
Liquid
Immunofluorescence, Immunocytochemistry, Western Blot
0.4 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O95229
ZWINT
ZWINT Fusion Protein Ag2933
20 μL
Primary
Human
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
ZWINT Rabbit anti-Human, Polyclonal, Proteintech > 20 μL; Unconjugated