missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zyxin Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10330-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
Zyxin binds alpha-actinin and the CRP proteins and is implicated in the regulation and modulation of actin filaments organization, cell adhesion, cell motility, mitosis, and signal transduction. Zyxin may shuttle between focal adhesion plaques and the nucleus. It has been suggested that Zyxin is involved in gene transcription by its association with transcription factors in the nucleus.Specifications
Zyxin | |
Polyclonal | |
Unconjugated | |
ZYX | |
ESP 2, HED 2, ZYX, zyxin, Zyxin 2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
7791 | |
-20°C | |
Liquid |
Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q15942 | |
ZYX | |
Zyxin Fusion Protein Ag0419 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |