missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zyxin Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | Zyxin |
---|---|
Concentration | 0.13 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16826653
|
Proteintech
10330-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 17-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16816653
|
Proteintech
10330-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 17-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Zyxin binds alpha-actinin and the CRP proteins and is implicated in the regulation and modulation of actin filaments organization, cell adhesion, cell motility, mitosis, and signal transduction. Zyxin may shuttle between focal adhesion plaques and the nucleus. It has been suggested that Zyxin is involved in gene transcription by its association with transcription factors in the nucleus.Specifications
Zyxin | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q15942 | |
7791 | |
Zyxin Fusion Protein Ag0419 | |
Primary | |
-20°C | |
ZYX |
0.13 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
ESP 2, HED 2, ZYX, zyxin, Zyxin 2 | |
ZYX | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |