CAS RN 10379-11-0
CAS RN 10379-11-0
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
2
av
2
Resultat
1
7-Chloro-5-(cyclohex-1-enyl)-1,3-dihydro-2H-1,4-benzodiazepin-2-one (Nortetrazepam), Mikromol
Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
Chlorphenamine Maleate (Chlorpheniramine Maleate) 1.0 mg/ml in Methanol (as free base), LoGiCal
LoGiCal delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.