CAS RN 1043932-31-5
CAS RN 1043932-31-5
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
3
av
3
Resultat
1
2-Chloropropanoic Acid 2-Methyl-4-thioxo-4H-pyran-3-yl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
2-Chloropropanoic Acid 2-Methyl-4-thioxo-4H-pyran-3-yl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
2-Chloropropanoic Acid 2-Methyl-4-thioxo-4H-pyran-3-yl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.