CAS RN 1044244-99-6
CAS RN 1044244-99-6
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
2
av
2
Resultat
1
Tetrahydro-Alpha-hydroxy-Alpha-methyl-2-(1-methylethoxy)-5-oxo-2H-pyran-4-acetic acid Methyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Tetrahydro-Alpha-hydroxy-Alpha-methyl-2-(1-methylethoxy)-5-oxo-2H-pyran-4-acetic acid Methyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.