CAS RN 104801-67-4
CAS RN 104801-67-4
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
2
av
2
Resultat
1
2-Hexyl-3-oxo-5-[(tetrahydro-2H-pyran-2-yl)oxy]-hexadecanoic Acid Methyl Ester(Mixture of Diastereomers), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
2-Hexyl-3-oxo-5-[(tetrahydro-2H-pyran-2-yl)oxy]-hexadecanoic Acid Methyl Ester(Mixture of Diastereomers), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.