CAS RN 105-65-7
CAS RN 105-65-7
IUPAC Namn:
(propan-2-yloxy)({[(propan-2-yloxy)methanethioyl]disulfanyl})methanethione
Synonymer:
thioperoxydicarbonic acid (((HO)C(S))2S2), bis(1
Molekylvikt (g/mol):
270.44
Molekylär sammansättning:
C8H14O2S4
InChi Key:
ZWWQICJTBOCQLA-UHFFFAOYSA-N
SMILES:
CC(C)OC(=S)SSC(=S)OC(C)C
1
–
3
av
3
Resultat
1
Diisopropylxanthogen Disulfide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Diisopropylxanthogen Disulfide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Diisopropylxanthogen Disulfide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.