CAS RN 10503-96-5
CAS RN 10503-96-5
IUPAC Namn:
1-(2-chloroethoxy)butane
Synonymer:
1-(2-chloroethoxy)butane
Molekylvikt (g/mol):
136.62
Molekylär sammansättning:
C6H13ClO
InChi Key:
OJMVRGXLTANFRG-UHFFFAOYSA-N
SMILES:
CCCCOCCCl
1
–
3
av
3
Resultat
1
1-(2-Chloroethoxy)butane, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
1-(2-Chloroethoxy)butane, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
1-(2-Chloroethoxy)butane, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.