CAS RN 105859-45-8
CAS RN 105859-45-8
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
3
av
3
Resultat
1
(2S)-3-{[tert-Butyl(dimethyl)silyl]oxy}-2-methylpropan-1-ol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(2S)-3-{[tert-Butyl(dimethyl)silyl]oxy}-2-methylpropan-1-ol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
(2S)-3-{[tert-Butyl(dimethyl)silyl]oxy}-2-methylpropan-1-ol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.