CAS RN 1072952-25-0
CAS RN 1072952-25-0
IUPAC Name:
[2-fluoro-5-(hydroxymethyl)phenyl]boronic acid
Synonyms:
2-fluoro-5-(hydroxymethyl)phenylboronic acid
Molecular Weight (g/mol):
169.95
Molecular Formula:
C7H8BFO3
InChi Key:
CMJGYVJNQJJJOZ-UHFFFAOYSA-N
SMILES:
OCC1=CC(B(O)O)=C(F)C=C1
1
–
1
of
1
results
1
Thermo Scientific Alfa Aesar 2-Fluoro-5-(hydroxymethyl)benzeneboronic acid, 98%, Thermo Scientific™
CAS: 1072952-25-0 Molecular Formula: C7H8BFO3 Molecular Weight (g/mol): 169.95 MDL Number: MFCD06656276 InChI Key: CMJGYVJNQJJJOZ-UHFFFAOYSA-N Synonym: 2-fluoro-5-hydroxymethylphenylboronic acid,2-fluoro-5-hydroxymethyl phenyl boronic acid,2-fluoro-5-hydroxymethyl phenylboronic acid,3-borono-4-fluorobenzyl alcohol,2-fluoro-5-hydroxymethylphenylbornic acid,2-fluoro-5-hydroxymethyl benzeneboronic acid,pubchem5040,acmc-2098uj,ksc494m6l PubChem CID: 44717544 IUPAC Name: [2-fluoro-5-(hydroxymethyl)phenyl]boronic acid SMILES: OCC1=CC(B(O)O)=C(F)C=C1