accessibility menu, dialog, popup

UserName

Discover Our 3 for 2 Promotion

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 107504-08-5

NFOHO

CAS RN 107504-08-5

IUPAC Name: 5-fluoropyridine-2-carboxylic acid
Synonyms: 5-fluoropyridine-2-carboxylic acid
Molecular Weight (g/mol): 141.1
Molecular Formula: C6H4FNO2
InChi Key: JTKFIIQGMVKDNZ-UHFFFAOYSA-N
SMILES: OC(=O)C1=NC=C(F)C=C1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products