accessibility menu, dialog, popup

UserName

Hard Science, Easy Bioreagents

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 108321-07-9

NNSOOOOHSOOOOHNaNa

CAS RN 108321-07-9

IUPAC Name: disodium 2-hydroxy-3-[4-(2-hydroxy-3-sulfonatopropyl)piperazin-1-yl]propane-1-sulfonate
Synonyms: disodium 2-hydroxy-3-[4-(2-hydroxy-3-sulfonatopropyl)piperazin-1-yl]propane-1-sulfonate
Molecular Weight (g/mol): 406.38
Molecular Formula: C10H20N2Na2O8S2
InChi Key: XMRCLLXGMGEVHS-UHFFFAOYNA-L
SMILES: [Na+].[Na+].OC(CN1CCN(CC(O)CS([O-])(=O)=O)CC1)CS([O-])(=O)=O
Quantity 
  • (1)
  • (1)
pH 
  • (2)

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products