accessibility menu, dialog, popup

UserName

Hard Science, Easy Bioreagents

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 108341-18-0

H2ONH2(R)OHOHHOOH(R)(R)OHHOOOHO

CAS RN 108341-18-0

IUPAC Name: (2R,3R)-2,3-dihydroxybutanedioic acid 4-[(1R)-2-amino-1-hydroxyethyl]benzene-1,2-diol hydrate
Synonyms: L(+)-tartaric acid norepinephrine hydrate
Molecular Weight (g/mol): 337.28
Molecular Formula: C12H19NO10
InChi Key: LNBCGLZYLJMGKP-LUDZCAPTSA-N
SMILES: O.O[C@H]([C@@H](O)C(O)=O)C(O)=O.NC[C@H](O)C1=CC=C(O)C(O)=C1
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products