accessibility menu, dialog, popup

UserName

Hard Science, Easy Bioreagents

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1087-02-1

CAS RN 1087-02-1

IUPAC Name: 1,4-dicyclohexylbenzene
Synonyms: 1,4-dicyclohexylbenzene
Molecular Weight (g/mol): 242.41
Molecular Formula: C18H26
InChi Key: QQFSIGWYINAJOB-UHFFFAOYSA-N
SMILES: C1CCC(CC1)C1=CC=C(C=C1)C1CCCCC1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products