accessibility menu, dialog, popup

UserName

Hard Science, Easy Bioreagents

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 108825-65-6

H3CNNNNNH3COCH3

CAS RN 108825-65-6

IUPAC Name: N-methyl-N-(3-{3-methyl-[1,2,4]triazolo[4,3-b]pyridazin-6-yl}phenyl)acetamide
Synonyms: N-methyl-N-(3-{3-methyl-[1,2,4]triazolo[4,3-b]pyridazin-6-yl}phenyl)acetamide
Molecular Weight (g/mol): 281.32
Molecular Formula: C15H15N5O
InChi Key: WPAQLESUVYGUJZ-UHFFFAOYSA-N
SMILES: CN(C(C)=O)C1=CC=CC(=C1)C1=NN2C(C)=NN=C2C=C1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products