CAS RN 108964-32-5
CAS RN 108964-32-5
IUPAC Name:
(acetyloxy)methyl 2-({2-[(acetyloxy)methoxy]-2-oxoethyl}[2-(5-{[(acetyloxy)methoxy]carbonyl}-1,3-oxazol-2-yl)-4-(2-{2-[bis({2-[(acetyloxy)methoxy]-2-oxoethyl})amino]-5-methylphenoxy}ethoxy)-1-benzofuran-5-yl]amino)acetate
Synonyms:
fura-2-acetoxymethyl ester
Molecular Weight (g/mol):
1001.86
Molecular Formula:
C44H47N3O24
InChi Key:
UTBYGIDPBQOHNT-UHFFFAOYSA-N
SMILES:
CC(=O)OCOC(=O)CN(CC(=O)OCOC(C)=O)C1=CC=C(C)C=C1OCCOC1=C2C=C(OC2=CC=C1N(CC(=O)OCOC(C)=O)CC(=O)OCOC(C)=O)C1=NC=C(O1)C(=O)OCOC(C)=O
1
–
5
of
5
results
5
MedChemExpress Fura-2 AM
MedChemExpress Fura-2 AM is a high affinity, intracellular, UV light-excitable and ratiometric fluorescent Ca2+ indicator.
