accessibility menu, dialog, popup

UserName

Nurture Your Progress - Savings to support your research

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 109-46-6

H3CHNSHNCH3

CAS RN 109-46-6

IUPAC Name: 1,3-dibutylthiourea
Synonyms: N,N'-di-n-butylthiourea
Molecular Weight (g/mol): 188.33
Molecular Formula: C9H20N2S
InChi Key: KFFQABQEJATQAT-UHFFFAOYSA-N
SMILES: CCCCNC(=S)NCCCC
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products