accessibility menu, dialog, popup

Test

CAS RN 11003-57-9

CAS RN 11003-57-9

IUPAC Name: 3,5-bis(3-amino-3-carboxypropyl)-4-(4-amino-4-carboxybutyl)-1-(5-amino-5-carboxypentyl)pyridin-1-ium
Synonyms: desmosine
Molecular Weight (g/mol): 526.61
Molecular Formula: C24H40N5O8
InChi Key: VEVRNHHLCPGNDU-UHFFFAOYNA-O
SMILES: NC(CCCC[N+]1=CC(CCC(N)C(O)=O)=C(CCCC(N)C(O)=O)C(CCC(N)C(O)=O)=C1)C(O)=O
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results