CAS RN 1100598-50-2
CAS RN 1100598-50-2
IUPAC Name:
5-bromo-2-[3-(chloromethyl)phenyl]pyrimidine
Synonyms:
5-bromo-2-[3-(chloromethyl)phenyl]pyrimidine
Molecular Weight (g/mol):
283.55
Molecular Formula:
C11H8BrClN2
InChi Key:
KHSZEUJOVUJWKR-UHFFFAOYSA-N
SMILES:
ClCC1=CC=CC(=C1)C1=NC=C(Br)C=N1
1
–
1
of
1
results
1
Thermo Scientific Alfa Aesar 5-Bromo-2-[3-(chloromethyl)phenyl]pyrimidine, 97%, Thermo Scientific™
CAS: 1100598-50-2 Molecular Formula: C11H8BrClN2 Molecular Weight (g/mol): 283.553 MDL Number: MFCD20527211 InChI Key: KHSZEUJOVUJWKR-UHFFFAOYSA-N Synonym: 5-bromo-2-3-chloromethyl phenyl pyrimidine,5-bromo-2-3-chloromethylphenyl pyrimidine,5-bromo-2-3-chloromethyl-phenyl-pyrimidine,pyrimidine, 5-bromo-2-3-chloromethyl phenyl PubChem CID: 59438047 IUPAC Name: 5-bromo-2-[3-(chloromethyl)phenyl]pyrimidine SMILES: C1=CC(=CC(=C1)CCl)C2=NC=C(C=N2)Br