CAS RN 1103926-82-4
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
4
av
4
Resultat
1
R&D Systems™ TM 5275 sodium salt
A Tocris product, part of R&D Systems. Plasminogen activator inhibitor-1 (PAI-1) inhibitor; orally bioavailable
2
TM5275 sodium, MedChemExpress
MedChemExpress TM5275 sodium is a plasminogen activator inhibitor (PAI-1) with an IC50 of 6.95 μM.
3
Tocris Bioscience™ TM 5275 sodium salt
Plasminogen activator inhibitor-1 (PAI-1) inhibitor; orally bioavailable
4
Tocris Bioscience™ TM 5275 sodium salt
Plasminogen activator inhibitor-1 (PAI-1) inhibitor; orally bioavailable