accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 110448-33-4

ISOONHN

CAS RN 110448-33-4

IUPAC Name: 1-[(5-iodonaphthalen-1-yl)sulfonyl]-1,4-diazepane
Synonyms: 1-(5-iodonaphthalen-1-ylsulfonyl)-1,4-diazepane
Molecular Weight (g/mol): 416.28
Molecular Formula: C15H17IN2O2S
InChi Key: GEHJIACZUFWBTK-UHFFFAOYSA-N
SMILES: IC1=C2C=CC=C(C2=CC=C1)S(=O)(=O)N1CCCNCC1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Grade 
  • (2)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products