accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 110683-10-8

H3COHNHOO(E)

CAS RN 110683-10-8

IUPAC Name: 2-[(2E)-3-(4-pentylphenyl)prop-2-enamido]benzoic acid
Synonyms: 2-[(2E)-3-(4-pentylphenyl)prop-2-enamido]benzoic acid
Molecular Weight (g/mol): 337.42
Molecular Formula: C21H23NO3
InChi Key: GAMRBCZMOOMBSQ-CCEZHUSRSA-N
SMILES: CCCCCC1=CC=C(\C=C\C(=O)NC2=CC=CC=C2C(O)=O)C=C1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products