CAS RN 11097-69-1
CAS RN 11097-69-1
IUPAC Name:
2,2',3,3',4-pentachloro-1,1'-biphenyl
Synonyms:
2,2',3,3',4-pentachloro-1,1'-biphenyl
Molecular Weight (g/mol):
326.42
Molecular Formula:
C12H5Cl5
InChi Key:
AUGNBQPSMWGAJE-UHFFFAOYSA-N
SMILES:
ClC1=CC=C(C(Cl)=C1Cl)C1=C(Cl)C(Cl)=CC=C1
1
–
9
of
9
results