accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 111-34-2

H3COCH2

CAS RN 111-34-2

IUPAC Name: 1-(ethenyloxy)butane
Synonyms: butyl vinyl ether
Molecular Weight (g/mol): 100.16
Molecular Formula: C6H12O
InChi Key: UZKWTJUDCOPSNM-UHFFFAOYSA-N
SMILES: CCCCOC=C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)
1x1 image pixel for data collection

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products