accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1112459-82-1

SNH2HCl

CAS RN 1112459-82-1

IUPAC Name: 1-{4-[(thiophen-2-yl)methyl]phenyl}methanamine hydrochloride
Synonyms: 1-[4-(thiophen-2-ylmethyl)phenyl]methanamine hydrochloride
Molecular Weight (g/mol): 239.76
Molecular Formula: C12H14ClNS
InChi Key: BRDMSBXGZSSVJT-UHFFFAOYSA-N
SMILES: Cl.NCC1=CC=C(CC2=CC=CS2)C=C1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products