accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1116-22-9

NH3(S)OHN(S)OOOOOO

CAS RN 1116-22-9

IUPAC Name: (2S)-2-[(4S)-4-azaniumyl-4-carboxylatobutanamido]pentanedioate
Synonyms: (2S)-2-[(4S)-4-ammonio-4-carboxylatobutanamido]pentanedioate
Molecular Weight (g/mol): 274.23
Molecular Formula: C10H14N2O7
InChi Key: OWQDWQKWSLFFFR-WDSKDSINSA-L
SMILES: [NH3+][C@@H](CCC(=O)N[C@@H](CCC([O-])=O)C([O-])=O)C([O-])=O
1x1 image pixel for data collection

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products