accessibility menu, dialog, popup

UserName

The Ultimate Lab Appliances Promotion

Save Up to 78%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 111652-20-1

CH3CH3H3COONHOOCH3CH3H3C

CAS RN 111652-20-1

IUPAC Name: tert-butyl 2-{[(tert-butoxy)carbonyl]amino}acetate
Synonyms: tert-butyl 2-[(tert-butoxycarbonyl)amino]acetate
Molecular Weight (g/mol): 231.29
Molecular Formula: C11H21NO4
InChi Key: ZDKFMHKWZATBMR-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)CNC(=O)OC(C)(C)C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products