accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 111771-08-5

OOFI

CAS RN 111771-08-5

IUPAC Name: 2-fluoro-6-iodobenzoate
Synonyms: 2-fluoro-6-iodobenzoate
Molecular Weight (g/mol): 265
Molecular Formula: C7H3FIO2
InChi Key: CYCXAPWOBWWNRK-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=C(F)C=CC=C1I
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products