accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 111865-47-5

H3CNH3CCH3HBrBrBr

CAS RN 111865-47-5

IUPAC Name: benzyltrimethylazanium dibromine hydrobromide
Synonyms: Brom benzyltrimethylazanium hydrobromide
Molecular Weight (g/mol): 390.96
Molecular Formula: C10H17Br3N
InChi Key: GSDDWZVWOXMLJZ-UHFFFAOYSA-N
SMILES: Br.BrBr.C[N+](C)(C)CC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products