accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1121-58-0

H3CNHN

CAS RN 1121-58-0

IUPAC Name: N-methylpyridin-4-amine
Synonyms: N-methylpyridin-4-amine
Molecular Weight (g/mol): 108.14
Molecular Formula: C6H8N2
InChi Key: LSCYTCMNCWMCQE-UHFFFAOYSA-N
SMILES: CNC1=CC=NC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products