accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1122-71-0

CH3OHN

CAS RN 1122-71-0

IUPAC Name: (6-methylpyridin-2-yl)methanol
Synonyms: (6-methylpyridin-2-yl)methanol
Molecular Weight (g/mol): 123.16
Molecular Formula: C7H9NO
InChi Key: JLVBSBMJQUMAMW-UHFFFAOYSA-N
SMILES: CC1=CC=CC(CO)=N1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products