accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1122-83-4

OSN

CAS RN 1122-83-4

IUPAC Name: N-(oxo-λ⁴-sulfanylidene)aniline
Synonyms: N-sulfinylaniline
Molecular Weight (g/mol): 139.17
Molecular Formula: C6H5NOS
InChi Key: FIOJWGRGPONADF-UHFFFAOYSA-N
SMILES: O=S=NC1=CC=CC=C1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products