CAS RN 112259-66-2
CAS RN 112259-66-2
IUPAC Name:
2-amino-4-(cyclohexyloxy)-4-oxobutanoic acid
Synonyms:
2-amino-4-(cyclohexyloxy)-4-oxobutanoic acid
Molecular Weight (g/mol):
215.25
Molecular Formula:
C10H17NO4
InChi Key:
IJAPBBOAGRLKMH-UHFFFAOYNA-N
SMILES:
NC(CC(=O)OC1CCCCC1)C(O)=O
1
–
1
of
1
results
1
Thermo Scientific Alfa Aesar L-Aspartic acid 4-cyclohexyl ester, 95%, Thermo Scientificâ„¢
CAS: 112259-66-2 Molecular Formula: C10H17NO4 Molecular Weight (g/mol): 215.25 MDL Number: MFCD00190762 InChI Key: IJAPBBOAGRLKMH-UHFFFAOYNA-N Synonym: h-asp ochex-oh,h-asp ochx-oh,l-aspartic acid beta-cyclohexyl ester,2s-2-amino-4-cyclohexyloxy-4-oxobutanoic acid,ambotzhaa5960,pubchem18997,l-aspartic acid b-cyclohexyl ester,h-asp ochex-oh hydrochloride salt PubChem CID: 14283862 IUPAC Name: 2-amino-4-(cyclohexyloxy)-4-oxobutanoic acid SMILES: NC(CC(=O)OC1CCCCC1)C(O)=O