accessibility menu, dialog, popup

UserName

Hard Science, Easy Bioreagents

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1124-63-6

HO

CAS RN 1124-63-6

IUPAC Name: 3-cyclohexylpropan-1-ol
Synonyms: cyclohexylpropanol
Molecular Weight (g/mol): 142.24
Molecular Formula: C9H18O
InChi Key: CLYAQFSQLQTVNO-UHFFFAOYSA-N
SMILES: OCCCC1CCCCC1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products